| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_001128080.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_001128080.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_001128081.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_001128081.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_001128082.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_001128082.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_020286.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
NM_020286.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700018806.mm.1 |
TC0700002087.mm.1 |
MTA TC |