Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277728_g1
          See other H2-M10.3 GE Assays ›
          Gene Symbol:
          H2-M10.3
          Gene Name:
          histocompatibility 2, M region locus 10.3
          Gene Aliases:
          5.3H
          Chromosome Location:
          Chr.17: 36365004 - 36368417 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277728_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-M10.3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201608.2 NP_963902.1 5 - 6 1004 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-M10.3

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          receptor binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129627 NM_201608.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129623 AY211080.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129623 NM_201608.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392903 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129627 NM_201608.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129623 AY211080.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129623 NM_201608.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98770 AY211080.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31163798 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08770662 - -
          Whole Mouse Genome G4122A A_52_P98770 AY211080.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116883 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13063457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13063439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31163819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13200343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13514666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13219671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31163839 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450775 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450774 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450772 - -
          Affymetrix Mouse Clariom S Array PSR1700019355.mm.1 NM_201608.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019355.mm.1 NM_201608.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019355.mm.1 TC1700002021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019357.mm.1 NM_201608.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019357.mm.1 NM_201608.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019357.mm.1 TC1700002021.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106450487 - -
          Illumina mouse-6 v1.0 expression beadchip 106450487 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260360 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247647 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248583 - -
          mouse-6 v1.1 (Illumina) GI_42475537-S - -
          Illumina Sentrix mouse-6.v1 bead array 106450487 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106450487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862866 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862866 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline