Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01278350_g1
          See other 6430531B16RIK GE Assays ›
          Gene Symbol:
          6430531B16Rik
          Gene Name:
          RIKEN cDNA 6430531B16 gene
          Gene Aliases:
          Gm1093
          Chromosome Location:
          Chr.7: 139972303 - 139978755 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01278350_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6430531B16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033465.2 NP_001028637.1 5 - 6 756 95
          GenBank mRNA AK131789.1 - 5 - 6 757 95
          BC145461.1 - 5 - 6 716 95
          BC145730.1 - 5 - 6 720 40131033 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004486.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700037628.mm.1 NM_001033465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037628.mm.1 NM_001033465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037628.mm.1 TC0700004486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037636.mm.1 NM_001033465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037636.mm.1 NM_001033465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037636.mm.1 TC0700004486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037634.mm.1 NM_001033465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037634.mm.1 NM_001033465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037634.mm.1 TC0700004486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037630.mm.1 NM_001033465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037630.mm.1 NM_001033465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037630.mm.1 TC0700004486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037631.mm.1 NM_001033465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037631.mm.1 NM_001033465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037631.mm.1 TC0700004486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037632.mm.1 NM_001033465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037632.mm.1 NM_001033465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037632.mm.1 TC0700004486.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037625.mm.1 NM_001033465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037625.mm.1 NM_001033465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037625.mm.1 TC0700004486.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 706757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline