Search
Search

Chaf1a| Gene Symbol: Chaf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013733.3 | NP_038761.1 | 1 - 2 | 136 | 75 | |
| GenBank mRNA | AK034839.1 | - | 1 - 2 | 137 | 75 | |
| AK135763.1 | - | 1 - 2 | 149 | 75 | ||
| BC053740.1 | - | 1 - 2 | 107 | 6504838 | 75 | |
Set Membership: |
|