Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Chaf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013733.3 | NP_038761.1 | 6 - 7 | 1335 | 68 | |
GenBank mRNA | AJ132771.2 | - | 6 - 7 | 1239 | 68 | |
AK034839.1 | - | 6 - 7 | 1336 | 68 | ||
AK161016.1 | - | 163 | 68 | |||
BC053740.1 | - | 6 - 7 | 1306 | 6504838 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |