Search
Search

Pot1b| Gene Symbol: Pot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028370.1 | NP_082646.1 | 3 - 4 | 211 | 88 | |
| GenBank mRNA | AK013364.1 | - | 3 - 4 | 212 | 88 | |
| AK045286.1 | - | 3 - 4 | 162 | 88 | ||
| AK050727.1 | - | 3 - 4 | 210 | 88 | ||
| AK165040.1 | - | 3 - 4 | 188 | 88 | ||
| BB642981.1 | - | 3 - 4 | 162 | 88 | ||
Set Membership: |
|