Search
Search
| Gene Symbol: Rgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009059.2 | NP_033085.2 | 13 - 14 | 1696 | 91 | |
| GenBank mRNA | AK144440.1 | - | 13 - 14 | 1698 | 91 | |
| BC068121.1 | - | 13 - 14 | 1696 | 6415928 | 91 | |
| KJ650210.1 | - | 12 - 13 | 1504 | 91 | ||
| U54639.1 | - | 13 - 14 | 1625 | 91 | ||
Set Membership: |
|