Search
Search

Wdr46| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020603.2 | NP_065628.1 | 12 - 13 | 1541 | 104 | |
| GenBank mRNA | BC013527.1 | - | 12 - 13 | 1584 | 104 | |
| BC026431.1 | - | 9 - 10 | 1165 | 104 | ||
| BC046977.1 | - | 12 - 13 | 1541 | 6315172 | 104 | |
| KJ650211.1 | - | 12 - 13 | 1526 | 104 | ||
| KJ663716.1 | - | 12 - 13 | 1394 | 104 | ||
Set Membership: |
|