Search
Search

4930430A15Rik| Gene Symbol: 4930430A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026248.3 | NP_080524.2 | 13 - 14 | 92 | ||
| GenBank mRNA | AK015247.1 | - | 13 - 14 | 1472 | 92 | |
| BC125324.1 | - | 13 - 14 | 40129839 | 92 | ||
Set Membership: |
|