Search
Search

Thbs2| Gene Symbol: Thbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011581.3 | NP_035711.2 | 12 - 13 | 2323 | 69 | |
| GenBank mRNA | AK132508.1 | - | 12 - 13 | 2154 | 69 | |
| BC031843.1 | - | 1 - 2 | 108 | 69 | ||
| BC053702.1 | - | 12 - 13 | 2125 | 30062369 | 69 | |
| L07803.1 | - | 12 - 13 | 2151 | 69 | ||
| M64866.1 | - | 12 - 13 | 2151 | 69 | ||
Set Membership: |
|