Search
Search

Smoc2| Gene Symbol: Smoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022315.2 | NP_071710.2 | 11 - 12 | 1493 | 90 | |
| GenBank mRNA | AJ249901.1 | - | 11 - 12 | 1464 | 87 | |
| AK030958.1 | - | 11 - 12 | 1462 | 90 | ||
| AK159749.1 | - | 11 - 12 | 1460 | 87 | ||
| AK160002.1 | - | 11 - 12 | 1460 | 87 | ||
Set Membership: |
|