Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01279258_m1
          See other ADGRF4 GE Assays ›
          Gene Symbol:
          Adgrf4
          Gene Name:
          adhesion G protein-coupled receptor F4
          Gene Aliases:
          4632435A09Rik, AV239010, Gpr115
          Chromosome Location:
          Chr.17: 42656887 - 42692284 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01279258_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adgrf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289499.1 NP_001276428.1 4 - 5 469 60
          NM_001289500.1 NP_001276429.1 4 - 5 456 60
          NM_001289501.1 NP_001276430.1 4 - 5 500 60
          NM_030067.2 NP_084343.1 4 - 5 543 60
          GenBank mRNA AK019508.1 - 4 - 5 546 60
          BC089564.1 - 4 - 5 469 30502915 60
          BC140980.1 - 4 - 5 409 9055592 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002114.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          signal transducer activity
          transmembrane signaling receptor activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700019792.mm.1 NM_001289499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019792.mm.1 NM_001289500.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019792.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019792.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019792.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019798.mm.1 NM_001289499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019798.mm.1 NM_001289500.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019798.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019798.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019798.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019801.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019801.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019801.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019793.mm.1 NM_001289499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019793.mm.1 NM_001289500.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019793.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019793.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019793.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019797.mm.1 NM_001289499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019797.mm.1 NM_001289500.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019797.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019797.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019797.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019805.mm.1 NM_001289499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019805.mm.1 NM_001289500.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019805.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019805.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019805.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019794.mm.1 NM_001289499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019794.mm.1 NM_001289500.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019794.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019794.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019794.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019804.mm.1 NM_001289499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019804.mm.1 NM_001289500.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019804.mm.1 NM_001289501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019804.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019804.mm.1 TC1700002114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019800.mm.1 NM_030067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019800.mm.1 TC1700002114.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0