Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01280106_m1
          See other 1700122O11RIK GE Assays ›
          Gene Symbol:
          1700122O11Rik
          Gene Name:
          RIKEN cDNA 1700122O11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 48036745 - 48038293 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01280106_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700122O11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029689.1 NP_083965.1 1 - 2 78 89
          GenBank mRNA AK007241.1 - 1 - 2 82 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002221.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139096 AK007241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P139096 AK007241.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109562 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139096 AK007241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P139096 AK007241.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109562 - -
          Whole Mouse Genome G4122A A_51_P139096 AK007241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P139096 AK007241.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13380696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13391213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13380691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13251306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13154309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13268710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13136371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13380693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13124339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13364025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13154305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13154310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13391211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13583334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31215566 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432496_PM_at - -
          Mouse Genome 430 2.0 Array 1432496_at - -
          Mouse Genome 430 2.0 Array 1432496_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76651_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76651_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700020988.mm.1 NM_029689.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020988.mm.1 NM_029689.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020988.mm.1 TC1700002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020990.mm.1 NM_029689.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020990.mm.1 NM_029689.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020990.mm.1 TC1700002221.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020989.mm.1 NM_029689.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020989.mm.1 NM_029689.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020989.mm.1 TC1700002221.mm.1 MTA TC
          Mouse Expression Set 430 1432496_PM_at - -
          Mouse Expression Set 430 1432496_at - -
          Affymetrix HT Mouse Genome 430B Array 1432496_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432496_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774131 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4560170 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline