Search
Search

Olfr472| Gene Symbol: Olfr472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146774.1 | NP_666985.1 | 1 | 263 | 103 | |
| GenBank mRNA | BC138079.1 | - | 1 | 355 | 8861098 | 103 |
| BC145756.1 | - | 1 | 355 | 40131059 | 103 | |
Set Membership: |
|