Search
Search

Rbm45| Gene Symbol: Rbm45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153405.2 | NP_700454.1 | 7 - 8 | 1218 | 70 | |
| GenBank mRNA | AB036992.1 | - | 7 - 8 | 1134 | 70 | |
| AK080420.1 | - | 7 - 8 | 1192 | 70 | ||
| AK089975.1 | - | 7 - 8 | 1111 | 70 | ||
| AK090084.1 | - | 7 - 8 | 1187 | 70 | ||
| AK140861.1 | - | 7 - 8 | 1219 | 70 | ||
| AY152391.1 | - | 7 - 8 | 1218 | 70 | ||
| BC057890.1 | - | 7 - 8 | 1129 | 4979982 | 70 | |
| CB597326.1 | - | 1 - 2 | 63 | 70 | ||
Set Membership: |
|