Search
Search

Nphp1| Gene Symbol: Nphp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291012.1 | NP_001277941.1 | 6 - 7 | 675 | 89 | |
| NM_001291013.1 | NP_001277942.1 | 6 - 7 | 675 | 89 | ||
| NM_016902.4 | NP_058598.1 | 6 - 7 | 675 | 89 | ||
| GenBank mRNA | AF127180.1 | - | 6 - 7 | 638 | 89 | |
| AF176210.1 | - | 6 - 7 | 638 | 89 | ||
| AK159627.1 | - | 6 - 7 | 652 | 89 | ||
| BC118953.2 | - | 6 - 7 | 638 | 40093235 | 89 | |
| EF174553.3 | - | 6 - 7 | 647 | 89 | ||
Set Membership: |
|