Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01282122_mH
          See other VMN2R123 GE Assays ›
          Gene Symbol:
          Vmn2r123
          Gene Name:
          vomeronasal 2, receptor 123
          Gene Aliases:
          V2r1
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01282122_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r123
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009485.1 NP_033511.1 4 - 5 1581 65
          GenBank mRNA AF011411.1 - 4 - 5 1581 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002112.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn2r123

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981756 AF011418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981756 AF011419.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981756 NR_028141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981756 NR_038052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981756 AF011418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981756 AF011419.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981756 NR_028141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981756 NR_038052.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120466 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116274 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116297 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414574 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598118 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606559 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414561 - -
          Affymetrix Mouse Clariom S Array PSR0400017857.mm.1 NR_028141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017857.mm.1 NR_028141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017857.mm.1 NR_038052.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017857.mm.1 TC0400002112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400017856.mm.1 NR_028141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017856.mm.1 NR_028141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017856.mm.1 NR_038052.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017856.mm.1 TC0400002112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400017854.mm.1 TC0400002112.mm.1 MTA TC
          Murine Genome U74v2 Set 95798_f_at - -
          Murine Genome U74v2 Set 95797_PM_f_at - -
          Murine Genome U74v2 Set 100677_PM_f_at - -
          Murine Genome U74v2 Set 100677_f_at - -
          Murine Genome U74v2 Set 95798_PM_f_at - -
          Murine Genome U74v2 Set 101644_PM_f_at - -
          Murine Genome U74v2 Set 101630_PM_f_at - -
          Murine Genome U74v2 Set 101630_f_at - -
          Murine Genome U74v2 Set 101644_f_at - -
          Murine Genome U74v2 Set 95797_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2421137 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2458220 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2500445 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2421137 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2500445 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2458220 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline