| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200002231.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002231.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002231.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002255.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002255.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002255.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002239.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002239.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002239.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002248.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002248.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002248.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002245.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002245.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002245.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002246.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002246.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002246.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002249.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002249.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002249.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002250.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002250.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002250.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002256.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002256.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002256.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002240.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002240.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002240.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002230.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002230.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002230.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002252.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002252.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002252.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002254.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002254.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002254.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002244.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002244.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002244.mm.1 |
TC1200000287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200002253.mm.1 |
NM_001033436.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002253.mm.1 |
NM_001033436.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200002253.mm.1 |
TC1200000287.mm.1 |
MTA TC |