Search
Search

Ppp1r12b| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081307.1 | NP_001074776.1 | 10 - 11 | 1615 | 124 | |
| GenBank mRNA | AK141375.1 | - | 10 - 11 | 1604 | 124 | |
| BC145746.1 | - | 10 - 11 | 1526 | 40131049 | 124 | |
Set Membership: |
|