| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100011638.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011638.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011638.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011638.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011638.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011632.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011632.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011632.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011632.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011632.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011633.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011633.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011633.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011633.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011633.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011635.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011635.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011635.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011635.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011635.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011636.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011636.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011636.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011636.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011636.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011637.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011637.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011637.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011637.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011637.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011631.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011631.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011631.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011631.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011631.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011630.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011630.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011630.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011630.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011630.mm.1 |
TC0100001419.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011634.mm.1 |
NM_001136104.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011634.mm.1 |
NM_001136104.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011634.mm.1 |
NM_009595.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011634.mm.1 |
NM_009595.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011634.mm.1 |
TC0100001419.mm.1 |
MTA TC |