Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01286916_s1
          See other DNAJB3 GE Assays ›
          Gene Symbol:
          Dnajb3
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily B, member 3
          Gene Aliases:
          Hsj3, MSJ-1, Msj1
          Chromosome Location:
          Chr.1: 88204732 - 88205748 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01286916_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008299.3 NP_032325.2 1 962 74
          GenBank mRNA AK005690.1 - 1 911 74
          U95607.1 - 1 964 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajb3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422168 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422163 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422161 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422162 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422166 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422165 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422167 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976237 NM_008299.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976237 AK005690.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147570 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976237 NM_008299.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976237 AK005690.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20422186 - -
          Whole Mouse Genome G4122A A_51_P501840 NM_008299.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501840 AK005690.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501840 U95607.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00651420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00557109 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00557108 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422163 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422161 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422168 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422167 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422162 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422166 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20422165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20422166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01132032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00557109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00557093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00646774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00646768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00646763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00651419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20422162 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10356460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356458 - -
          Affymetrix HT MG-430 PM Array Plate 1423553_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10356457 - -
          Mouse Genome 430 2.0 Array 1423553_PM_at - -
          Mouse Genome 430 2.0 Array 1423553_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15504_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15504_PM_at - -
          Mouse Expression Set 430 1423553_at - -
          Mouse Expression Set 430 1423553_PM_at - -
          Murine Genome U74v2 Set 97480_PM_f_at - -
          Murine Genome U74v2 Set 97481_r_at - -
          Murine Genome U74v2 Set 97481_PM_r_at - -
          Murine Genome U74v2 Set 97480_f_at - -
          Murine Genome U74v2 Set 169315_r_at - -
          Murine Genome U74v2 Set 169315_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1423553_PM_at - -
          Mouse Genome 430A 2.0 Array 1423553_at - -
          Affymetrix HT Mouse Genome 430A Array 1423553_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423553_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 379387 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470128 - -
          Illumina mouse-6 v1.0 expression beadchip 2470128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770825 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2770825 - -
          mouse-6 v1.1 (Illumina) scl16502.1.45_71-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16502.1.45_71-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2869728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2869728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770825 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline