Search
Search
| Gene Symbol: Sag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009118.2 | NP_033144.1 | 10 - 11 | 1163 | 99 | |
| GenBank mRNA | AK044214.1 | - | 9 - 10 | 844 | 99 | |
| AK053278.1 | - | 10 - 11 | 922 | 99 | ||
| AK143098.1 | - | 10 - 11 | 922 | 99 | ||
| AK149299.1 | - | 10 - 11 | 1172 | 99 | ||
| BC016498.1 | - | 10 - 11 | 956 | 4527551 | 99 | |
| M24086.1 | - | 10 - 11 | 950 | 99 | ||
Set Membership: |
|