Search
Search

Lrrcc1| Gene Symbol: Lrrcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163579.1 | NP_001157051.1 | 11 - 12 | 1893 | 83 | |
| NM_001163580.1 | NP_001157052.1 | 11 - 12 | 1845 | 83 | ||
| NM_028915.3 | NP_083191.2 | 11 - 12 | 1893 | 83 | ||
| GenBank mRNA | AK004646.1 | - | 11 - 12 | 1894 | 83 | |
| AK016559.1 | - | 11 - 12 | 1832 | 83 | ||
| AK029563.1 | - | 11 - 12 | 1811 | 83 | ||
| AK030800.1 | - | 3 - 4 | 487 | 83 | ||
| AK173256.1 | - | 11 - 12 | 1821 | 83 | ||
| BC061202.1 | - | 11 - 12 | 1801 | 83 | ||
| BC138734.1 | - | 11 - 12 | 1867 | 8861756 | 83 | |
| BC138735.1 | - | 11 - 12 | 1867 | 8861757 | 83 | |
Set Membership: |
|