Search
Search

Rapgef6| Gene Symbol: Rapgef6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252494.1 | NP_001239423.1 | 24 - 25 | 3829 | 73 | |
| NM_001252496.1 | NP_001239425.1 | 25 - 26 | 3853 | 73 | ||
| NM_175258.4 | NP_780467.2 | 23 - 24 | 3814 | 73 | ||
| GenBank mRNA | AK143331.1 | - | 11 - 12 | 2201 | 73 | |
| AK220459.1 | - | 13 - 14 | 2241 | 73 | ||
| BC019702.1 | - | 2 - 3 | 391 | 73 | ||
| BC025553.1 | - | 6 - 7 | 1308 | 73 | ||
| BC059847.1 | - | 6 - 7 | 1234 | 73 | ||
| BC092525.1 | - | 6 - 7 | 1381 | 73 | ||
| BC141182.1 | - | 24 - 25 | 3728 | 9055794 | 73 | |
| BC145503.1 | - | 25 - 26 | 3753 | 73 | ||
Set Membership: |
|