Search
Search

Mapk9| Gene Symbol: Mapk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163672.1 | NP_001157144.1 | 6 - 7 | 891 | 64 | |
| NM_207692.2 | NP_997575.2 | 6 - 7 | 891 | 64 | ||
| GenBank mRNA | AF052466.1 | - | 5 - 6 | 627 | 64 | |
| AF052467.1 | - | 5 - 6 | 627 | 64 | ||
| AK031959.1 | - | 6 - 7 | 890 | 64 | ||
| AK090057.1 | - | 6 - 7 | 835 | 64 | ||
| AK145083.1 | - | 6 - 7 | 866 | 64 | ||
| BC028341.1 | - | 6 - 7 | 873 | 4192796 | 64 | |
Set Membership: |
|