Search
Search

Sqstm1| Gene Symbol: Sqstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290769.1 | NP_001277698.1 | 2 - 3 | 365 | 110 | |
| NM_011018.3 | NP_035148.1 | 2 - 3 | 365 | 110 | ||
| GenBank mRNA | BC006019.1 | - | 2 - 3 | 362 | 3487289 | 110 |
| E08146.1 | - | 2 - 3 | 323 | 110 | ||
| U40930.1 | - | 2 - 3 | 335 | 110 | ||
| U57413.1 | - | 2 - 3 | 327 | 110 | ||
Set Membership: |
|