Search
Search

Clint1| Gene Symbol: Clint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045520.3 | NP_001038985.2 | 8 - 9 | 1198 | 84 | |
| GenBank mRNA | AK143544.1 | - | 8 - 9 | 1199 | 84 | |
| AK147875.1 | - | 8 - 9 | 1183 | 84 | ||
| AK168991.1 | - | 8 - 9 | 1164 | 84 | ||
| BC004080.1 | - | 5 - 6 | 736 | 84 | ||
Set Membership: |
|