Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01289320_m1
          See other MTFR1 GE Assays ›
          Gene Symbol:
          Mtfr1
          Gene Name:
          mitochondrial fission regulator 1
          Gene Aliases:
          1300002C08Rik, 1700080D04Rik, 2810026O10Rik, 4930579E05Rik, Chppr, Fam54a2
          Chromosome Location:
          Chr.3: 19187326 - 19220817 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01289320_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtfr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253390.1 NP_001240319.1 6 - 7 891 94
          NM_001253391.1 NP_001240320.1 6 - 7 1053 94
          NM_026182.5 NP_080458.1 6 - 7 901 94
          NR_045575.1 7 - 8 938 94
          GenBank mRNA AF354708.1 - 5 - 6 852 94
          AK004846.1 - 6 - 7 879 94
          AK006952.1 - 6 - 7 1053 94
          AK009195.2 - 6 - 7 866 94
          AK012823.1 - 879 94
          AK016316.1 - 6 - 7 1041 94
          AK075822.1 - 6 - 7 877 94
          AK075992.1 - 6 - 7 901 94
          AK134822.1 - 7 - 8 895 94
          AK161251.1 - 6 - 7 874 94
          BC019971.1 - 6 - 7 849 4159093 94
          BC042471.1 - 5 - 6 825 3988195 94
          DQ369744.1 - 6 - 7 1012 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000124.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mtfr1

          Gene Ontology Categories:

          Function(s)

          mitochondrial fission
          aerobic respiration

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NM_001253390.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NM_001253390.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NM_001253391.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NM_001253391.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NM_026182.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NM_026182.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NR_045575.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 NR_045575.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000536.mm.1 TC0300000124.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 931424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline