| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300000847.mm.1 |
NM_001164056.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000847.mm.1 |
NM_001164056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000847.mm.1 |
NM_008875.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000847.mm.1 |
NM_008875.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000847.mm.1 |
TC0300000168.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000848.mm.1 |
NM_001164056.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000848.mm.1 |
NM_001164056.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000848.mm.1 |
NM_008875.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000848.mm.1 |
NM_008875.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000848.mm.1 |
TC0300000168.mm.1 |
MTA TC |