Search
Search

Hspa4l| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011020.3 | NP_035150.3 | 14 - 15 | 1879 | 143 | |
| GenBank mRNA | AB001926.1 | - | 14 - 15 | 1845 | 143 | |
| AK033950.1 | - | 1 - 2 | 108 | 143 | ||
| AK144225.1 | - | 14 - 15 | 1916 | 143 | ||
| BC012712.1 | - | 14 - 15 | 1948 | 4190161 | 143 | |
| BC057002.1 | - | 14 - 15 | 1879 | 5702602 | 143 | |
| BC110662.1 | - | 14 - 15 | 1899 | 4511154 | 143 | |
| D49482.1 | - | 14 - 15 | 1873 | 143 | ||
| U23921.1 | - | 14 - 15 | 1996 | 143 | ||
Set Membership: |
|