Search
Search

Mfsd4| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114662.1 | NP_001108134.1 | 4 - 5 | 1015 | 86 | |
| NM_172510.4 | NP_766098.2 | 4 - 5 | 1015 | 86 | ||
| GenBank mRNA | AK038882.1 | - | 4 - 5 | 908 | 86 | |
| AK044667.1 | - | 4 - 5 | 888 | 86 | ||
| AK048692.1 | - | 3 - 4 | 817 | 86 | ||
| AK162495.1 | - | 4 - 5 | 919 | 86 | ||
| BC058790.1 | - | 4 - 5 | 864 | 6395070 | 86 | |
Set Membership: |
|