Search
Search

Dusp27| Gene Symbol: Dusp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033344.3 | NP_001028516.2 | 3 - 4 | 340 | 77 | |
| NM_001160049.1 | NP_001153521.1 | 3 - 4 | 336 | 77 | ||
| GenBank mRNA | AK048603.1 | - | 3 - 4 | 363 | 77 | |
| AK142263.1 | - | 3 - 4 | 341 | 77 | ||
| BC117936.1 | - | 3 - 4 | 317 | 40097789 | 77 | |
| BY730343.1 | - | 3 - 4 | 341 | 77 | ||
Set Membership: |
|