Search
Search

Frem2| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172862.3 | NP_766450.2 | 7 - 8 | 6405 | 71 | |
| GenBank mRNA | AB236662.1 | - | 7 - 8 | 6405 | 71 | |
| AJ833643.1 | - | 7 - 8 | 6134 | 71 | ||
| CN530725.1 | - | 2 - 3 | 291 | 71 | ||
Set Membership: |
|