Search
Search
| Gene Symbol: Ahr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013464.4 | NP_038492.1 | 6 - 7 | 1053 | 87 | |
| GenBank mRNA | AF405560.1 | - | 6 - 7 | 687 | 87 | |
| AF405561.1 | - | 6 - 7 | 687 | 87 | ||
| AF405562.1 | - | 6 - 7 | 687 | 87 | ||
| AF405563.1 | - | 6 - 7 | 687 | 87 | ||
| AF405565.1 | - | 6 - 7 | 687 | 87 | ||
| AF405566.1 | - | 6 - 7 | 687 | 87 | ||
| AF405567.1 | - | 6 - 7 | 687 | 87 | ||
| AF405568.1 | - | 6 - 7 | 687 | 87 | ||
| AF405570.1 | - | 6 - 7 | 687 | 87 | ||
| AK153678.1 | - | 6 - 7 | 1054 | 87 | ||
| AK155380.1 | - | 6 - 7 | 1105 | 87 | ||
| AY662080.1 | - | 6 - 7 | 687 | 87 | ||
| AY662081.1 | - | 5 - 6 | 583 | 87 | ||
| AY662082.1 | - | 6 - 7 | 87 | |||
| AY662083.1 | - | 6 - 7 | 687 | 87 | ||
| D38416.1 | - | 6 - 7 | 702 | 87 | ||
Set Membership: |
|