Search
Search

Ccna2| Gene Symbol: Ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009828.2 | NP_033958.2 | 5 - 6 | 1143 | 72 | |
| GenBank mRNA | AK044924.1 | - | 5 - 6 | 1146 | 72 | |
| AK135861.1 | - | 5 - 6 | 1115 | 72 | ||
| AK182616.1 | - | 1 - 2 | 126 | 72 | ||
| AK187939.1 | - | 1 - 2 | 109 | 72 | ||
| AK204936.1 | - | 1 - 2 | 61 | 72 | ||
| BC052730.1 | - | 5 - 6 | 1091 | 6816226 | 72 | |
| X75483.1 | - | 5 - 6 | 1091 | 72 | ||
| Z26580.1 | - | 5 - 6 | 1100 | 72 | ||
Set Membership: |
|