Search
Search

Soat1| Gene Symbol: Soat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009230.3 | NP_033256.2 | 11 - 12 | 1292 | 66 | |
| GenBank mRNA | AK052299.1 | - | 8 - 9 | 849 | 66 | |
| AK142486.1 | - | 11 - 12 | 1209 | 66 | ||
| AK159393.1 | - | 11 - 12 | 1222 | 66 | ||
| AK159529.1 | - | 11 - 12 | 1222 | 66 | ||
| AK160017.1 | - | 11 - 12 | 1222 | 66 | ||
| BC083092.1 | - | 10 - 11 | 1106 | 30446741 | 66 | |
| DQ903181.1 | - | 11 - 12 | 1209 | 66 | ||
| S81092.1 | - | 11 - 12 | 1226 | 66 | ||
Set Membership: |
|