Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292838_s1
          See other CLEC14A GE Assays ›
          Gene Symbol:
          Clec14a
          Gene Name:
          C-type lectin domain family 14, member a
          Gene Aliases:
          1200003C23Rik, AI642649
          Chromosome Location:
          Chr.12: 58264720 - 58269258 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292838_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec14a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025809.5 NP_080085.3 1 4403 117
          GenBank mRNA AK045596.1 - 1 4032 117
          BB307353.2 - 1 554 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec14a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155977 AK045596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155977 AK080201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155977 NM_025809.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155977 AK045596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155977 AK080201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155977 NM_025809.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999082 AK164665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999082 BC019452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999082 AK033733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999082 AK045596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999082 AK080201.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654718 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142766 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999082 AK164665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999082 BC019452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999082 AK033733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999082 AK045596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999082 AK080201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155977 AK045596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155977 AK080201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155977 NM_025809.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155977 AK045596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155977 AK080201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155977 NM_025809.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28466492 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142766 - -
          Whole Mouse Genome G4122A A_52_P566665 AK033733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566665 AK045596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566665 AK080201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566665 NM_025809.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566665 AK004557.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155977 AK045596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155977 AK080201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155977 NM_025809.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155977 AK045596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155977 AK080201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155977 NM_025809.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129026 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142766 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336549 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336565 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336552 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10336564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28466456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10096395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10336546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10336557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10336555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09885133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10150704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10392890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10336534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10336537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10096404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28466533 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10400514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400513 - -
          Affymetrix HT MG-430 PM Array Plate 1419468_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419467_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10400510 - -
          Mouse Genome 430 2.0 Array 1419467_PM_at - -
          Mouse Genome 430 2.0 Array 1419467_at - -
          Mouse Genome 430 2.0 Array 1419468_PM_at - -
          Mouse Genome 430 2.0 Array 1419468_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66864_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66864_PM_at - -
          Mouse Expression Set 430 1419468_PM_at - -
          Mouse Expression Set 430 1419467_PM_at - -
          Mouse Expression Set 430 1419468_at - -
          Mouse Expression Set 430 1419467_at - -
          Mouse Genome 430A 2.0 Array 1419468_PM_at - -
          Mouse Genome 430A 2.0 Array 1419468_at - -
          Mouse Genome 430A 2.0 Array 1419467_at - -
          Mouse Genome 430A 2.0 Array 1419467_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419468_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419467_at - -
          Affymetrix HT Mouse Genome 430A Array 1419467_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419468_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3610673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256550 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256550 - -
          mouse-6 v1.1 (Illumina) scl42426.2_100-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42426.2_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256550 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256550 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline