Search
Search
| Gene Symbol: Cpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029706.1 | NP_083982.1 | 8 - 9 | 849 | 68 | |
| GenBank mRNA | AK003061.1 | - | 8 - 9 | 786 | 68 | |
| AK008678.1 | - | 5 - 6 | 521 | 68 | ||
| BC138759.1 | - | 8 - 9 | 807 | 8861781 | 68 | |
| BC138762.1 | - | 8 - 9 | 807 | 8861784 | 68 | |
Set Membership: |
|