Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01293117_m1
          See other GLI2 GE Assays ›
          Gene Symbol:
          Gli2
          Gene Name:
          GLI-Kruppel family member GLI2
          Gene Aliases:
          AW546128
          Chromosome Location:
          Chr.1: 118834061 - 119053619 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01293117_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gli2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081125.1 NP_001074594.1 8 - 9 1523 62
          GenBank mRNA AM261668.1 - 5 - 6 748 62
          AM261669.1 - 5 - 6 767 62
          AM261670.1 - 6 - 7 854 62
          AM262741.1 - 6 - 7 805 62
          AM262742.1 - 7 - 8 1040 62
          AM262743.1 - 7 - 8 1137 62
          AM262744.1 - 9 - 10 1354 62
          BC082604.1 - 8 - 9 1464 30672479 62
          BC085190.1 - 8 - 9 1439 30694633 62
          X99104.1 - 8 - 9 1283 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002942.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          osteoblast differentiation
          in utero embryonic development
          kidney development
          morphogenesis of an epithelium
          chondrocyte differentiation
          osteoblast development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          smoothened signaling pathway
          pattern specification process
          axon guidance
          ventral midline development
          hindgut morphogenesis
          heart development
          cell proliferation
          positive regulation of cell proliferation
          regulation of smoothened signaling pathway
          epidermal cell differentiation
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          floor plate formation
          spinal cord dorsal/ventral patterning
          ventral spinal cord development
          spinal cord motor neuron differentiation
          cerebellar cortex morphogenesis
          smoothened signaling pathway involved in ventral spinal cord interneuron specification
          smoothened signaling pathway involved in spinal cord motor neuron cell fate specification
          dorsal/ventral neural tube patterning
          neural tube development
          smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
          spinal cord ventral commissure morphogenesis
          pituitary gland development
          cell differentiation
          lung development
          mammary gland development
          hindbrain development
          negative regulation of chondrocyte differentiation
          tube development
          odontogenesis of dentin-containing tooth
          embryonic digit morphogenesis
          positive regulation of neuron differentiation
          positive regulation of DNA replication
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic digestive tract development
          developmental growth
          anatomical structure formation involved in morphogenesis
          neuron development
          branching morphogenesis of an epithelial tube
          anatomical structure development
          notochord regression
          head development
          prostatic bud formation
          mammary gland duct morphogenesis
          smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          cellular response to organic cyclic compound
          cochlea morphogenesis
          cellular response to virus
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          nucleic acid binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          zinc ion binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100023625.mm.1 NM_001081125.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023625.mm.1 NM_001081125.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023625.mm.1 TC0100002942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023627.mm.1 NM_001081125.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023627.mm.1 NM_001081125.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023627.mm.1 TC0100002942.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline