Search
Search

Lrrc34| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027941.1 | NP_082217.1 | 7 - 8 | 766 | 62 | |
| GenBank mRNA | AK005720.1 | - | 7 - 8 | 818 | 62 | |
| BC100519.1 | - | 7 - 8 | 766 | 30919449 | 62 | |
| BC116419.1 | - | 6 - 7 | 599 | 62 | ||
| BC116420.1 | - | 6 - 7 | 606 | 40103399 | 62 | |
| JF934738.1 | - | 7 - 8 | 717 | 62 | ||
Set Membership: |
|