Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295389_g1
          See other DCAF6 GE Assays ›
          Gene Symbol:
          Dcaf6
          Gene Name:
          DDB1 and CUL4 associated factor 6
          Gene Aliases:
          1200006M05Rik, Iqwd1, PC326
          Chromosome Location:
          Chr.1: 165329500 - 165460553 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295389_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dcaf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028759.1 NP_083035.1 18 - 19 2801 72
          GenBank mRNA AK004618.1 - 18 - 19 2664 72
          AK077238.1 - 9 - 10 1319 72
          AK161893.1 - 6 - 7 2535 72
          BC058980.1 - 16 - 17 2364 72
          BC084732.1 - 16 - 17 2364 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transcription from RNA polymerase II promoter
          ligand-dependent nuclear receptor transcription coactivator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 BC084732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 AK161893.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 BC058980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 AK077238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 NM_028759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 BC084732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 AK161893.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 BC058980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 AK077238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140797 NM_028759.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212322 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516403 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724743 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485415 AK007964.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485415 AK032497.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485415 AK135054.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485415 AY445814.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485415 NM_144807.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452215 AK013225.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452215 NM_181392.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159433 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134908 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122991 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 BC084732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 AK161893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 BC058980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 AK077238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 NM_028759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 BC084732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 AK161893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 BC058980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 AK077238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140797 NM_028759.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340123 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134908 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340206 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340065 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340261 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122991 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00240534 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340046 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340274 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340179 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340100 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04340148 - -
          Whole Mouse Genome G4122A A_51_P140797 BC084732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 AK161893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 BC058980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 AK077238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 NM_028759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 BC084732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 AK161893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 BC058980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 AK077238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P140797 NM_028759.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134908 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122991 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04340065 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04340123 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04340261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01075440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00190862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00146985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00978335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00137229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20794100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00978338 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10359739 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359740 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359735 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359738 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359737 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359736 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359742 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359741 - -
          Affymetrix HT MG-430 PM Array Plate 1447049_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453312_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10359734 - -
          Mouse Genome 430 2.0 Array 1447049_at - -
          Mouse Genome 430 2.0 Array 1453312_at - -
          Mouse Genome 430 2.0 Array 1447049_PM_at - -
          Mouse Genome 430 2.0 Array 1453312_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74106_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74106_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100028237.mm.1 NM_028759.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028237.mm.1 NM_028759.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028237.mm.1 TC0100003443.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028238.mm.1 NM_028759.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028238.mm.1 NM_028759.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028238.mm.1 TC0100003443.mm.1 MTA TC
          Mouse Expression Set 430 1447049_PM_at - -
          Mouse Expression Set 430 1447049_at - -
          Mouse Expression Set 430 1453312_PM_at - -
          Mouse Expression Set 430 1453312_at - -
          Murine Genome U74v2 Set 131136_PM_at - -
          Murine Genome U74v2 Set 131136_at - -
          Affymetrix HT Mouse Genome 430B Array 1453312_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453312_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 301143 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2608666 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2608666 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline