Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008371.4 | NP_032397.1 | 3 - 4 | 478 | 135 | |
GenBank mRNA | AK040399.1 | - | 3 - 4 | 262 | 135 | |
AK041307.1 | - | 3 - 4 | 479 | 135 | ||
AK052452.1 | - | 4 - 5 | 1166 | 135 | ||
AK139858.1 | - | 4 - 5 | 839 | 135 | ||
AK145184.1 | - | 3 - 4 | 769 | 135 | ||
BC110553.2 | - | 3 - 4 | 387 | 40047584 | 135 | |
X07962.1 | - | 3 - 4 | 778 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |