Search
Search
| Gene Symbol: Pkp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019645.2 | NP_062619.1 | 8 - 9 | 1516 | 69 | |
| GenBank mRNA | AK036856.1 | - | 8 - 9 | 1701 | 69 | |
| AK079429.1 | - | 8 - 9 | 1701 | 69 | ||
| BC139041.1 | - | 8 - 9 | 1611 | 8862063 | 69 | |
| BC139042.1 | - | 8 - 9 | 1612 | 8862064 | 69 | |
| Y07941.1 | - | 8 - 9 | 1516 | 69 | ||
Set Membership: |
|