Search
Search
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 8 - 9 | 953 | 87 | |
| GenBank mRNA | AK034591.1 | - | 8 - 9 | 848 | 87 | |
| AK079849.1 | - | 8 - 9 | 890 | 87 | ||
| AK083401.1 | - | 8 - 9 | 867 | 87 | ||
| AK083537.1 | - | 8 - 9 | 874 | 87 | ||
| AK087134.1 | - | 8 - 9 | 929 | 87 | ||
| AK134406.1 | - | 8 - 9 | 844 | 87 | ||
| AK165243.1 | - | 8 - 9 | 868 | 87 | ||
| AK166118.1 | - | 8 - 9 | 868 | 87 | ||
| AK169523.1 | - | 8 - 9 | 905 | 87 | ||
| BC018191.1 | - | 8 - 9 | 854 | 3994248 | 87 | |
| BC025501.1 | - | 8 - 9 | 900 | 5321013 | 87 | |
| BC026604.1 | - | 7 - 8 | 1407 | 87 | ||
Set Membership: |
|