Search
Search
| Gene Symbol: Tns4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172564.3 | NP_766152.2 | 8 - 9 | 1778 | 82 | |
| GenBank mRNA | AK036834.1 | - | 8 - 9 | 1775 | 82 | |
| AK039855.1 | - | 3 - 4 | 404 | 82 | ||
| AK170638.1 | - | 2 - 3 | 150 | 82 | ||
| BC055820.1 | - | 8 - 9 | 1760 | 4209161 | 82 | |
Set Membership: |
|