Search
Search

Rapgefl1| Gene Symbol: Rapgefl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080925.1 | NP_001074394.1 | 6 - 7 | 1486 | 81 | |
| GenBank mRNA | BC080279.1 | - | 5 - 6 | 554 | 81 | |
| BC130266.1 | - | 7 - 8 | 762 | 40143028 | 81 | |
Set Membership: |
|