Search
Search

Tbc1d22b| Gene Symbol: Tbc1d22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198647.1 | NP_941049.1 | 10 - 11 | 1307 | 73 | |
| GenBank mRNA | BC045600.1 | - | 10 - 11 | 1307 | 4489206 | 73 |
| BC060066.1 | - | 5 - 6 | 579 | 73 | ||
Set Membership: |
|