| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100012220.mm.1 |
NM_001166642.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012220.mm.1 |
NM_001166642.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012220.mm.1 |
NM_001166643.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012220.mm.1 |
NM_001166643.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012220.mm.1 |
NM_138681.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012220.mm.1 |
NM_138681.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012220.mm.1 |
TC1100001293.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012217.mm.1 |
NM_001166642.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012217.mm.1 |
NM_001166642.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012217.mm.1 |
NM_138681.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012217.mm.1 |
NM_138681.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012217.mm.1 |
TC1100001293.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012222.mm.1 |
NM_001166642.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012222.mm.1 |
NM_001166642.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012222.mm.1 |
NM_001166643.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012222.mm.1 |
NM_001166643.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012222.mm.1 |
NM_138681.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012222.mm.1 |
NM_138681.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012222.mm.1 |
TC1100001293.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012214.mm.1 |
NM_001166642.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012214.mm.1 |
NM_001166642.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012214.mm.1 |
NM_001166643.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012214.mm.1 |
NM_001166643.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012214.mm.1 |
NM_138681.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012214.mm.1 |
NM_138681.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012214.mm.1 |
TC1100001293.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012219.mm.1 |
NM_001166642.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012219.mm.1 |
NM_001166642.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012219.mm.1 |
NM_001166643.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012219.mm.1 |
NM_001166643.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012219.mm.1 |
NM_138681.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012219.mm.1 |
NM_138681.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012219.mm.1 |
TC1100001293.mm.1 |
MTA TC |