Search
Search

Rictor| Gene Symbol: Rictor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030168.3 | NP_084444.3 | 19 - 20 | 1735 | 99 | |
| GenBank mRNA | AK036149.1 | - | 19 - 20 | 1736 | 99 | |
| AK082586.1 | - | 19 - 20 | 1726 | 99 | ||
| AY497009.1 | - | 19 - 20 | 1713 | 99 | ||
| AY540053.1 | - | 19 - 20 | 1733 | 99 | ||
| BC022906.1 | - | 14 - 15 | 1281 | 99 | ||
| BC120903.1 | - | 15 - 16 | 1374 | 40086992 | 99 | |
| BC120904.1 | - | 19 - 20 | 1733 | 40086995 | 99 | |
Set Membership: |
|