Search
Search

Heatr6| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145432.3 | NP_663407.2 | 4 - 5 | 668 | 73 | |
| GenBank mRNA | AK012216.1 | - | 4 - 5 | 612 | 73 | |
| AK076219.1 | - | 4 - 5 | 796 | 73 | ||
| AK132494.1 | - | 4 - 5 | 603 | 73 | ||
| AK149059.1 | - | 4 - 5 | 462 | 73 | ||
| AK158323.1 | - | 4 - 5 | 798 | 73 | ||
| AK169901.1 | - | 4 - 5 | 615 | 73 | ||
| BC065092.1 | - | 4 - 5 | 594 | 6855410 | 73 | |
Set Membership: |
|